siRNA Details
| Gene Symbol: Scamp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100636.1 | NP_001094106.1 | 9 | 1028 | ||
| GenBank mRNA | BC090322.1 | - | 6 | 695 | ||
| L22079.1 | - | 9 | 948 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scamp1| Gene Symbol: Scamp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100636.1 | NP_001094106.1 | 9 | 1028 | ||
| GenBank mRNA | BC090322.1 | - | 6 | 695 | ||
| L22079.1 | - | 9 | 948 | |||