siRNA Details
| Gene Symbol: Pla2g1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031585.1 | NP_113773.1 | 3 | 291 | ||
| XM_008769222.1 | XP_008767444.1 | 3 | 399 | |||
| GenBank mRNA | D00036.1 | - | 3 | 291 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pla2g1b| Gene Symbol: Pla2g1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031585.1 | NP_113773.1 | 3 | 291 | ||
| XM_008769222.1 | XP_008767444.1 | 3 | 399 | |||
| GenBank mRNA | D00036.1 | - | 3 | 291 | ||