siRNA Details
| Gene Symbol: Ptgs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017232.3 | NP_058928.3 | 2 | 103 | ||
| GenBank mRNA | AF233596.1 | - | 227 | |||
| AY157736.1 | - | 2 | 103 | |||
| L20085.1 | - | 2 | 103 | |||
| L25925.1 | - | 2 | 109 | |||
| S67722.1 | - | 227 | ||||
| U03389.1 | - | 227 | ||||
| U04300.1 | - | 2 | 103 | |||

Ptgs2
Genomic Map