siRNA Details
| Gene Symbol: Ighmbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031586.1 | NP_113774.1 | 5 | 670 | ||
| GenBank mRNA | AF199411.1 | - | 5 | 670 | ||
| BC099790.1 | - | 5 | 866 | 7934952 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ighmbp2| Gene Symbol: Ighmbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031586.1 | NP_113774.1 | 5 | 670 | ||
| GenBank mRNA | AF199411.1 | - | 5 | 670 | ||
| BC099790.1 | - | 5 | 866 | 7934952 | ||