siRNA Details
| Gene Symbol: Pebp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017236.1 | NP_058932.1 | 3 | 321 | ||
| GenBank mRNA | BC063171.1 | - | 3 | 338 | 6888051 | |
| FQ211637.1 | - | 3 | 282 | |||
| FQ212424.1 | - | 3 | 304 | |||
| FQ215504.1 | - | 3 | 332 | |||
| FQ224857.1 | - | 3 | 332 | |||
| X71873.1 | - | 3 | 296 | |||
| X75253.1 | - | 3 | 321 | |||

Pebp1
Genomic Map