siRNA Details
| Gene Symbol: Lcn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024136.1 | NP_077050.1 | 5 | 498 | ||
| XM_008761580.1 | XP_008759802.1 | 5 | 1563 | |||
| GenBank mRNA | M12790.1 | - | 5 | 450 | ||
| X59832.1 | - | 5 | 490 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lcn5| Gene Symbol: Lcn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024136.1 | NP_077050.1 | 5 | 498 | ||
| XM_008761580.1 | XP_008759802.1 | 5 | 1563 | |||
| GenBank mRNA | M12790.1 | - | 5 | 450 | ||
| X59832.1 | - | 5 | 490 | |||