siRNA Details
| Gene Symbol: Pdha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004072.2 | NP_001004072.2 | 2 | 143 | ||
| GenBank mRNA | BC079369.1 | - | 2 | 135 | ||
| BC098897.1 | - | 2 | 143 | 7103543 | ||
| Z12158.1 | - | 2 | 162 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdha1| Gene Symbol: Pdha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004072.2 | NP_001004072.2 | 2 | 143 | ||
| GenBank mRNA | BC079369.1 | - | 2 | 135 | ||
| BC098897.1 | - | 2 | 143 | 7103543 | ||
| Z12158.1 | - | 2 | 162 | |||