siRNA Details
| Gene Symbol: Vipr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017238.1 | NP_058934.1 | 10 | 1070 | ||
| GenBank mRNA | U09631.1 | - | 10 | 1070 | ||
| Z25885.1 | - | 10 | 1023 | |||
More Information
Other Species Targeted
Set Membership: |
|

Vipr2
Genomic Map