siRNA Details
| Gene Symbol: Myh6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017239.2 | NP_058935.2 | 10 | 959 | ||
| GenBank mRNA | X15938.1 | - | 10 | 956 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myh6| Gene Symbol: Myh6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017239.2 | NP_058935.2 | 10 | 959 | ||
| GenBank mRNA | X15938.1 | - | 10 | 956 | ||