siRNA Details
Gene Symbol: Myh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017240.2 | NP_058936.1 | 14 | 1854 | ||
XM_006251951.2 | XP_006252013.1 | 16 | 2004 | |||
GenBank mRNA | X15939.1 | - | 14 | 1856 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Myh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017240.2 | NP_058936.1 | 14 | 1854 | ||
XM_006251951.2 | XP_006252013.1 | 16 | 2004 | |||
GenBank mRNA | X15939.1 | - | 14 | 1856 |