siRNA Details
| Gene Symbol: Fcgrt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033351.2 | NP_203502.1 | 3 | 680 | ||
| XM_006229043.1 | XP_006229105.1 | 4 | 789 | |||
| XM_008759343.1 | XP_008757565.1 | 4 | 688 | |||
| GenBank mRNA | BC061975.1 | - | 4 | 632 | 5623787 | |
| M35495.1 | - | 3 | 682 | |||
| X14323.1 | - | 3 | 682 | |||

Fcgrt
Genomic Map