siRNA Details
| Gene Symbol: Hif1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024359.1 | NP_077335.1 | 10 | 1319 | ||
| XM_006240196.2 | XP_006240258.1 | 10 | 1632 | |||
| XM_006240197.2 | XP_006240259.1 | 10 | 1629 | |||
| XM_006240198.2 | XP_006240260.1 | 10 | 1628 | |||
| XM_006240199.2 | XP_006240261.1 | 10 | 1331 | |||
| XM_008764695.1 | XP_008762917.1 | 10 | 1416 | |||
| GenBank mRNA | AF057308.1 | - | 10 | 1319 | ||
| Y09507.1 | - | 10 | 1416 | |||

Hif1a
Genomic Map