siRNA Details
| Gene Symbol: Lsamp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017242.1 | NP_058938.1 | 6 | 909 | ||
| XM_006248339.2 | XP_006248401.1 | 6 | 1360 | |||
| GenBank mRNA | AY326256.1 | - | 6 | 878 | ||
| BC087607.1 | - | 6 | 880 | 7314244 | ||
| U31554.1 | - | 6 | 909 | |||

Lsamp
Genomic Map