siRNA Details
| Gene Symbol: Eef2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017245.2 | NP_058941.1 | 2 | 133 | ||
| GenBank mRNA | BC066661.1 | - | 2 | 133 | 5623322 | |
| Y07504.1 | - | 2 | 119 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eef2| Gene Symbol: Eef2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017245.2 | NP_058941.1 | 2 | 133 | ||
| GenBank mRNA | BC066661.1 | - | 2 | 133 | 5623322 | |
| Y07504.1 | - | 2 | 119 | |||