siRNA Details
| Gene Symbol: Pcolce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019237.1 | NP_062110.1 | 6 | 1011 | ||
| GenBank mRNA | AB008534.1 | - | 6 | 998 | ||
| AF016503.1 | - | 6 | 1005 | |||
| BC086572.1 | - | 7 | 1067 | 7304047 | ||
| FQ209381.1 | - | 6 | 1049 | |||
| U94710.1 | - | 6 | 1011 | |||

Pcolce
Genomic Map