siRNA Details
| Gene Symbol: Pcolce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019237.1 | NP_062110.1 | 8 | 1268 | ||
| GenBank mRNA | AB008534.1 | - | 8 | 1255 | ||
| AF016503.1 | - | 8 | 1262 | |||
| BC086572.1 | - | 9 | 1324 | 7304047 | ||
| FQ209381.1 | - | 8 | 1306 | |||
| U94710.1 | - | 8 | 1268 | |||

Pcolce
Genomic Map