siRNA Details
Gene Symbol: Scn10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017247.1 | NP_058943.1 | 14 | 2119 | ||
XM_008766673.1 | XP_008764895.1 | 14 | 2167 | |||
GenBank mRNA | X92184.1 | - | 14 | 2119 |
