siRNA Details
| Gene Symbol: Esyt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017249.2 | NP_058945.2 | 10 | 1418 | ||
| GenBank mRNA | AF099138.1 | - | 10 | 1363 | ||
| BC101857.1 | - | 10 | 1418 | 7937926 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Esyt1| Gene Symbol: Esyt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017249.2 | NP_058945.2 | 10 | 1418 | ||
| GenBank mRNA | AF099138.1 | - | 10 | 1363 | ||
| BC101857.1 | - | 10 | 1418 | 7937926 | ||