siRNA Details
| Gene Symbol: Fdft1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019238.2 | NP_062111.1 | 5 | 732 | ||
| XM_008770746.1 | XP_008768968.1 | 4 | 3379 | |||
| GenBank mRNA | BC081810.1 | - | 5 | 732 | 7188993 | |
| FQ213323.1 | - | 5 | 715 | |||
| FQ219372.1 | - | 5 | 701 | |||
| M95591.1 | - | 5 | 750 | |||

Fdft1
Genomic Map