siRNA Details
| Gene Symbol: Htr2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017250.1 | NP_058946.1 | 1 | 443 | ||
| XM_006245369.2 | XP_006245431.1 | 1 | 2146 | |||
| XM_008767216.1 | XP_008765438.1 | 2 | 1293 | |||
| GenBank mRNA | X66842.1 | - | 1 | 443 | ||
More Information
Set Membership: |
|

Htr2b
Genomic Map