siRNA Details
| Gene Symbol: Gjb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019240.1 | NP_062113.1 | 1 | 618 | ||
| XM_006238845.2 | XP_006238907.1 | 2 | 1928 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gjb3| Gene Symbol: Gjb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019240.1 | NP_062113.1 | 1 | 618 | ||
| XM_006238845.2 | XP_006238907.1 | 2 | 1928 | |||