siRNA Details
| Gene Symbol: Bcat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017253.2 | NP_058949.1 | 1, 2 | 146 | ||
| GenBank mRNA | BC087710.1 | - | 1, 2 | 146 | 7315507 | |
| U35774.1 | - | 1, 2 | 112 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bcat1| Gene Symbol: Bcat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017253.2 | NP_058949.1 | 1, 2 | 146 | ||
| GenBank mRNA | BC087710.1 | - | 1, 2 | 146 | 7315507 | |
| U35774.1 | - | 1, 2 | 112 | |||