siRNA Details
Gene Symbol: Htr2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017254.1 | NP_058950.1 | 3 | 1421 | ||
GenBank mRNA | M30705.1 | - | 3 | 1421 | ||
M64867.1 | - | 1827 | ||||
X13971.1 | - | 1964 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
