siRNA Details
| Gene Symbol: Atp2b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133288.1 | NP_579822.1 | 23 | 4246 | ||
| XM_008773619.1 | XP_008771841.1 | 21 | 3590 | |||
| XM_008773620.1 | XP_008771842.1 | 20 | 3548 | |||
| XM_008773621.1 | XP_008771843.1 | 20 | 3503 | |||
| XM_008773622.1 | XP_008771844.1 | 19 | 3461 | |||
| XM_008773623.1 | XP_008771845.1 | 21 | 3657 | |||
| XM_008773627.1 | XP_008771849.1 | 20 | 3560 | |||
| GenBank mRNA | BC089969.1 | - | 5 | 727 | ||
| J05087.1 | - | 23 | 4246 | |||

Atp2b3
Genomic Map