siRNA Details
| Gene Symbol: Atp2b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133288.1 | NP_579822.1 | 14 | 2766 | ||
| XM_008773619.1 | XP_008771841.1 | 12 | 2177 | |||
| XM_008773620.1 | XP_008771842.1 | 11 | 2135 | |||
| XM_008773621.1 | XP_008771843.1 | 12 | 2177 | |||
| XM_008773622.1 | XP_008771844.1 | 11 | 2135 | |||
| XM_008773623.1 | XP_008771845.1 | 12 | 2177 | |||
| XM_008773624.1 | XP_008771846.1 | 12 | 2177 | |||
| XM_008773625.1 | XP_008771847.1 | 12 | 2177 | |||
| XM_008773626.1 | XP_008771848.1 | 12 | 2177 | |||
| XM_008773627.1 | XP_008771849.1 | 12 | 2234 | |||
| GenBank mRNA | J05087.1 | - | 14 | 2766 | ||

Atp2b3
Genomic Map