siRNA Details
| Gene Symbol: Ptgfrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019243.1 | NP_062116.1 | 5 | 1403 | ||
| GenBank mRNA | U26595.1 | - | 5 | 1403 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptgfrn| Gene Symbol: Ptgfrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019243.1 | NP_062116.1 | 5 | 1403 | ||
| GenBank mRNA | U26595.1 | - | 5 | 1403 | ||