siRNA Details
| Gene Symbol: Pitx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019247.1 | NP_062120.1 | 4 | 574 | ||
| XM_006231478.2 | XP_006231540.1 | 4 | 753 | |||
| GenBank mRNA | AJ011005.2 | - | 4 | 574 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pitx3| Gene Symbol: Pitx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019247.1 | NP_062120.1 | 4 | 574 | ||
| XM_006231478.2 | XP_006231540.1 | 4 | 753 | |||
| GenBank mRNA | AJ011005.2 | - | 4 | 574 | ||