siRNA Details
Gene Symbol: Tgfbr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017256.1 | NP_058952.1 | 6 | 980 | ||
GenBank mRNA | M77809.1 | - | 6 | 980 | ||
M80784.1 | - | 6 | 965 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tgfbr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017256.1 | NP_058952.1 | 6 | 980 | ||
GenBank mRNA | M77809.1 | - | 6 | 980 | ||
M80784.1 | - | 6 | 965 |