siRNA Details
Gene Symbol: Dbt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053312.1 | NP_445764.1 | 7 | 893 | ||
GenBank mRNA | BC101904.1 | - | 8 | 1105 | ||
BC166487.1 | - | 7 | 880 | 7103882 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dbt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053312.1 | NP_445764.1 | 7 | 893 | ||
GenBank mRNA | BC101904.1 | - | 8 | 1105 | ||
BC166487.1 | - | 7 | 880 | 7103882 |