siRNA Details
| Gene Symbol: Dbt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053312.1 | NP_445764.1 | 9, 10 | 1231 | ||
| GenBank mRNA | BC101904.1 | - | 10, 11 | 1443 | ||
| BC166487.1 | - | 9, 10 | 1218 | 7103882 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dbt| Gene Symbol: Dbt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053312.1 | NP_445764.1 | 9, 10 | 1231 | ||
| GenBank mRNA | BC101904.1 | - | 10, 11 | 1443 | ||
| BC166487.1 | - | 9, 10 | 1218 | 7103882 | ||