siRNA Details
| Gene Symbol: Ralgds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019250.2 | NP_062123.2 | 9 | 1675 | ||
| XM_006233750.2 | XP_006233812.1 | 9 | 2220 | |||
| XM_006233751.2 | XP_006233813.1 | 9 | 4153 | |||
| XM_006233752.2 | XP_006233814.1 | 9 | 1733 | |||
| XM_006233753.2 | XP_006233815.1 | 9 | 1697 | |||
| XM_008761632.1 | XP_008759854.1 | 9 | 1998 | |||
| GenBank mRNA | CB775259.1 | - | 3 | 347 | ||
| L07925.1 | - | 9 | 1675 | |||

Ralgds
Genomic Map