siRNA Details
| Gene Symbol: F2rl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053313.1 | NP_445765.1 | 2 | 1014 | ||
| GenBank mRNA | AF310076.1 | - | 2 | 1014 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

F2rl2| Gene Symbol: F2rl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053313.1 | NP_445765.1 | 2 | 1014 | ||
| GenBank mRNA | AF310076.1 | - | 2 | 1014 | ||