siRNA Details
| Gene Symbol: Hmgcs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017268.1 | NP_058964.1 | 10 | 1435 | ||
| XM_006231959.1 | XP_006232021.1 | 9 | 1604 | |||
| GenBank mRNA | AY724522.1 | - | 9 | 1421 | ||
| FQ211444.1 | - | 9 | 1407 | |||
| X52625.1 | - | 10 | 1435 | |||

Hmgcs1
Genomic Map