siRNA Details
| Gene Symbol: Hmgcs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017268.1 | NP_058964.1 | 5 | 693 | ||
| XM_006231959.1 | XP_006232021.1 | 4 | 862 | |||
| GenBank mRNA | AY724522.1 | - | 4 | 679 | ||
| FQ211444.1 | - | 4 | 665 | |||
| X52625.1 | - | 5 | 693 | |||

Hmgcs1
Genomic Map