siRNA Details
| Gene Symbol: Adam7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020301.1 | NP_064697.1 | 6 | 597 | ||
| GenBank mRNA | X66140.1 | - | 6 | 597 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adam7| Gene Symbol: Adam7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020301.1 | NP_064697.1 | 6 | 597 | ||
| GenBank mRNA | X66140.1 | - | 6 | 597 | ||