siRNA Details
| Gene Symbol: Ptpn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019253.3 | NP_062126.3 | 9 | 1498 | ||
| XM_006229252.2 | XP_006229314.1 | 14 | 2403 | |||
| XM_006229253.2 | XP_006229315.1 | 14 | 2344 | |||
| XM_006229254.2 | XP_006229316.1 | 13 | 2243 | |||
| XM_006229255.2 | XP_006229317.1 | 13 | 2319 | |||
| XM_006229257.2 | XP_006229319.1 | 12 | 2150 | |||
| XM_008759346.1 | XP_008757568.1 | 14 | 1878 | |||
| GenBank mRNA | BC092196.1 | - | 7 | 863 | 7313460 | |
| FQ211673.1 | - | 14 | 2007 | |||
| FQ213308.1 | - | 11 | 1312 | |||
| S49400.1 | - | 9 | 1498 | |||

Ptpn5
Genomic Map