siRNA Details
| Gene Symbol: Ptpn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019253.3 | NP_062126.3 | 5, 6 | 1145 | ||
| XM_006229252.2 | XP_006229314.1 | 10, 11 | 2050 | |||
| XM_006229253.2 | XP_006229315.1 | 10, 11 | 1991 | |||
| XM_006229254.2 | XP_006229316.1 | 9, 10 | 1890 | |||
| XM_006229255.2 | XP_006229317.1 | 9, 10 | 1966 | |||
| XM_006229257.2 | XP_006229319.1 | 8, 9 | 1797 | |||
| XM_008759346.1 | XP_008757568.1 | 10, 11 | 1525 | |||
| GenBank mRNA | BC092196.1 | - | 3, 4 | 510 | 7313460 | |
| FQ211673.1 | - | 10, 11 | 1654 | |||
| FQ213308.1 | - | 7, 8 | 959 | |||
| S49400.1 | - | 5, 6 | 1145 | |||

Ptpn5
Genomic Map