siRNA Details
| Gene Symbol: Cacng1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019255.1 | NP_062128.1 | 4 | 534 | ||
| GenBank mRNA | FQ215004.1 | - | 4 | 631 | ||
| FQ215755.1 | - | 4 | 613 | |||
| FQ216734.1 | - | 4 | 649 | |||
| FQ224130.1 | - | 4 | 705 | |||
| Y09453.1 | - | 4 | 534 | |||

Cacng1
Genomic Map