siRNA Details
| Gene Symbol: P2rx7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019256.1 | NP_062129.1 | 10 | 1148 | ||
| XM_008769223.1 | XP_008767445.1 | 10 | 1322 | |||
| GenBank mRNA | FJ436445.1 | - | 10 | 1186 | ||
| X95882.1 | - | 10 | 1148 | |||

P2rx7
Genomic Map