siRNA Details
| Gene Symbol: Psma2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017279.1 | NP_058975.1 | 8 | 616 | ||
| GenBank mRNA | BC060576.1 | - | 8 | 635 | 6919255 | |
| FQ214776.1 | - | 8 | 631 | |||
| FQ223872.1 | - | 631 | ||||
| J02897.1 | - | 8 | 616 | |||

Psma2
Genomic Map