siRNA Details
Gene Symbol: Psma3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017280.2 | NP_058976.1 | 11 | 1584 | ||
GenBank mRNA | BC081817.1 | - | 11 | 1584 | 7110009 | |
FQ224319.1 | - | 1611 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Psma3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017280.2 | NP_058976.1 | 11 | 1584 | ||
GenBank mRNA | BC081817.1 | - | 11 | 1584 | 7110009 | |
FQ224319.1 | - | 1611 |