siRNA Details
| Gene Symbol: Psmb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017284.1 | NP_058980.1 | 2 | 236 | ||
| GenBank mRNA | BC058487.1 | - | 2 | 256 | 6891616 | |
| D21799.1 | - | 2 | 236 | |||
| FQ217737.1 | - | 2 | 266 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psmb2| Gene Symbol: Psmb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017284.1 | NP_058980.1 | 2 | 236 | ||
| GenBank mRNA | BC058487.1 | - | 2 | 256 | 6891616 | |
| D21799.1 | - | 2 | 236 | |||
| FQ217737.1 | - | 2 | 266 | |||