siRNA Details
| Gene Symbol: Psmb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017284.1 | NP_058980.1 | 1 | 134 | ||
| GenBank mRNA | BC058487.1 | - | 1 | 154 | 6891616 | |
| D21799.1 | - | 1 | 134 | |||
| FQ217737.1 | - | 1 | 164 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_011970, BC008265, CB321399, AK164463, BB853639, and others.

Psmb2
Genomic Map