siRNA Details
| Gene Symbol: C1qb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019262.1 | NP_062135.1 | 3 | 629 | ||
| GenBank mRNA | FQ227589.1 | - | 3 | 554 | ||
| FQ228182.1 | - | 3 | 489 | |||
| FQ234810.1 | - | 3 | 568 | |||
| X71127.1 | - | 3 | 629 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C1qb| Gene Symbol: C1qb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019262.1 | NP_062135.1 | 3 | 629 | ||
| GenBank mRNA | FQ227589.1 | - | 3 | 554 | ||
| FQ228182.1 | - | 3 | 489 | |||
| FQ234810.1 | - | 3 | 568 | |||
| X71127.1 | - | 3 | 629 | |||