siRNA Details
| Gene Symbol: Pde1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022710.1 | NP_073201.1 | 4, 5 | 489 | ||
| XM_006242386.2 | XP_006242448.1 | 3, 4 | 1061 | |||
| GenBank mRNA | AF327906.1 | - | 4, 5 | 482 | ||
| FQ212460.1 | - | 4, 5 | 545 | |||
| M94537.1 | - | 4, 5 | 489 | |||

Pde1b
Genomic Map