siRNA Details
| Gene Symbol: Gabrr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017291.3 | NP_058987.2 | 3 | 310 | ||
| XM_008763609.1 | XP_008761831.1 | 3 | 93 | |||
| GenBank mRNA | U21070.1 | - | 3 | 267 | ||
| X95579.1 | - | 3 | 310 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gabrr1| Gene Symbol: Gabrr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017291.3 | NP_058987.2 | 3 | 310 | ||
| XM_008763609.1 | XP_008761831.1 | 3 | 93 | |||
| GenBank mRNA | U21070.1 | - | 3 | 267 | ||
| X95579.1 | - | 3 | 310 | |||