siRNA Details
| Gene Symbol: Prkar2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019264.2 | NP_062137.1 | 7, 8 | 901 | ||
| XM_006243712.2 | XP_006243774.1 | 8, 9 | 1086 | |||
| XM_006243713.2 | XP_006243775.1 | 8, 9 | 900 | |||
| XM_008766478.1 | XP_008764700.1 | 7, 8 | 851 | |||
| GenBank mRNA | AF533978.1 | - | 7, 8 | 905 | ||
| J02934.1 | - | 7, 8 | 699 | |||

Prkar2a
Genomic Map