siRNA Details
| Gene Symbol: Prkar2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019264.2 | NP_062137.1 | 7 | 854 | ||
| XM_006243712.2 | XP_006243774.1 | 8 | 1039 | |||
| XM_006243713.2 | XP_006243775.1 | 8 | 853 | |||
| XM_008766478.1 | XP_008764700.1 | 7 | 804 | |||
| GenBank mRNA | AF533978.1 | - | 7 | 858 | ||
| J02934.1 | - | 7 | 652 | |||

Prkar2a
Genomic Map