siRNA Details
| Gene Symbol: Gabra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183326.2 | NP_899155.1 | 8 | 980 | ||
| XM_006246123.2 | XP_006246185.1 | 8 | 972 | |||
| XM_006246124.2 | XP_006246186.1 | 8 | 805 | |||
| GenBank mRNA | AY574250.1 | - | 7 | 756 | ||
| BC100061.1 | - | 8 | 980 | 7122007 | ||
| FQ211937.1 | - | 8 | 1233 | |||

Gabra1
Genomic Map