siRNA Details
Gene Symbol: Gabra6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021841.1 | NP_068613.1 | 6 | 658 | ||
XM_006246128.2 | XP_006246190.1 | 6 | 807 | |||
XM_008767636.1 | XP_008765858.1 | 4 | 729 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gabra6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021841.1 | NP_068613.1 | 6 | 658 | ||
XM_006246128.2 | XP_006246190.1 | 6 | 807 | |||
XM_008767636.1 | XP_008765858.1 | 4 | 729 |